ENA423210350 Enamine

4-amino-8-methyl-1,2-dihydroquinoline-2-thione

  • MDL number MFCD20719270
  • Cas Number : 360078-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172185
Availability details for
Quantity
  • Availability
  • Add to Cart