ENA423210351 Enamine

5-[(4-methylphenyl)methyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD19347405
  • Cas Number : 14064-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173571
Availability details for
Quantity
  • Availability
  • Add to Cart