ENA423210354 Enamine

2-(propan-2-ylsulfanyl)-1H-imidazole

  • MDL number MFCD19219017
  • Cas Number : 7565-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173574
Availability details for
Quantity
  • Availability
  • Add to Cart