ENA423210363 Enamine

N-methyl-N-phenylaminosulfonamide

  • MDL number MFCD11205193
  • Cas Number : 127903-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173681

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-173681
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart