ENA423210366 Enamine

5-chloro-2-hydroxy-N-[3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD01538275
  • Cas Number : 1580-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173728
Availability details for
Quantity
  • Availability
  • Add to Cart