ENA423210383 Enamine

methyl 4-(2-bromoacetyl)benzoate

  • MDL number MFCD08062347
  • Cas Number : 56893-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23971
Availability details for
Quantity
  • Availability
  • Add to Cart