ENA423210385 Enamine

1-(4-acetylpiperazin-1-yl)-2-bromo-3-methylbutan-1-one

  • MDL number MFCD09931887
  • Cas Number : 1016752-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29147
Availability details for
Quantity
  • Availability
  • Add to Cart