ENA423210388 Enamine

2-(methylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD16041882
  • Cas Number : 1249519-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31346
Availability details for
Quantity
  • Availability
  • Add to Cart