ENA423210394 Enamine

[(3-methylphenyl)methyl](propan-2-yl)amine

  • MDL number MFCD08060640
  • Cas Number : 915922-51-9
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32100
Availability details for
Quantity
  • Availability
  • Add to Cart