ENA423210420 Enamine

[(2,4-difluorophenyl)methyl](propyl)amine

  • MDL number MFCD09928517
  • Cas Number : 637015-27-1
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32182
Availability details for
Quantity
  • Availability
  • Add to Cart