ENA423210463 Enamine

4-[(2-methylpropyl)amino]benzonitrile

  • MDL number MFCD11145736
  • Cas Number : 850570-33-1
  • Molecular Weight : 174.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32333
Availability details for
Quantity
  • Availability
  • Add to Cart