ENA423210482 Enamine

[(4-ethoxyphenyl)methyl](propan-2-yl)amine

  • MDL number MFCD08757432
  • Cas Number : 940196-09-8
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-ethoxybenzyl)-2-propanamine
Inchi Code 1S/C12H19NO/c1-4-14-12-7-5-11(6-8-12)9-13-10(2)3/h5-8,10,13H,4,9H2,1-3H3
Inchi Key ZDLALHMIFXHJAP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32413
Availability details for
Quantity
  • Availability
  • Add to Cart