ENA423210488 Enamine

[(2,4-dimethylphenyl)methyl](2-methoxyethyl)amine

  • MDL number MFCD11139308
  • Cas Number : 1019598-18-5
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32423
Availability details for
Quantity
  • Availability
  • Add to Cart