ENA423210495 Enamine

[(4-ethylphenyl)methyl](2-methoxyethyl)amine

  • MDL number MFCD07409064
  • Cas Number : 827328-12-1
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32443
Availability details for
Quantity
  • Availability
  • Add to Cart