ENA423210571 Enamine

[(2-fluorophenyl)methyl](2-methylpropyl)amine

  • MDL number MFCD04510051
  • Cas Number : 893589-79-2
  • Molecular Weight : 181.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32576

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-32576
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart