ENA423210587 Enamine

4-ethoxy-N-ethylaniline

  • MDL number MFCD03210680
  • Cas Number : 65570-13-0
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32669
Availability details for
Quantity
  • Availability
  • Add to Cart