ENA423210627 Enamine

butyl[(2,4-difluorophenyl)methyl]amine

  • MDL number MFCD11142429
  • Cas Number : 1019619-01-2
  • Molecular Weight : 199.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32758
Availability details for
Quantity
  • Availability
  • Add to Cart