ENA423210655 Enamine

(butan-2-yl)[2-(2-fluorophenyl)ethyl]amine

  • MDL number MFCD11143355
  • Cas Number : 1019530-90-5
  • Molecular Weight : 195.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32849
Availability details for
Quantity
  • Availability
  • Add to Cart