ENA423210722 Enamine

N-[(2-methylphenyl)methyl]cyclopropanamine

  • MDL number MFCD07408054
  • Cas Number : 625435-05-4
  • Molecular Weight : 161.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-33136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33136
Availability details for
Quantity
  • Availability
  • Add to Cart