ENA423210753 Enamine

2-[2-(dimethylamino)ethyl]aniline

  • MDL number MFCD09884768
  • Cas Number : 3478-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50928
Availability details for
Quantity
  • Availability
  • Add to Cart