ENA423210756 Enamine

1,3-dimethyl-1H,4H,5H,6H,7H-pyrazolo[3,4-b]pyridin-5-amine

  • MDL number MFCD09806704
  • Cas Number : 1016734-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53448
Availability details for
Quantity
  • Availability
  • Add to Cart