ENA423210758 Enamine

2-(2-bromoethoxy)-1,1-difluoroethane

  • MDL number MFCD24280786
  • Cas Number : 1544861-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-172209
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart