ENA423210761 Enamine

1-(7-fluoro-1-benzofuran-2-yl)ethan-1-one

  • MDL number MFCD11619469
  • Cas Number : 1094640-56-8
  • Molecular Weight : 178.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55085
Availability details for
Quantity
  • Availability
  • Add to Cart