ENA423210763 Enamine

2,2,2-trifluoroethyl N-[4-(propan-2-yl)-1,3-thiazol-2-yl]carbamate

  • MDL number MFCD21373486
  • Cas Number : 1477569-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56703
Availability details for
Quantity
  • Availability
  • Add to Cart