ENA423210773 Enamine

3-hydroxypentanenitrile

  • MDL number MFCD11654010
  • Cas Number : 13635-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70004
Availability details for
Quantity
  • Availability
  • Add to Cart