ENA423210785 Enamine

2-(5-chlorothiophen-2-yl)acetic acid

  • MDL number MFCD01859875
  • Cas Number : 13669-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84036
Availability details for
Quantity
  • Availability
  • Add to Cart