ENA423210793 Enamine

3-(4-methyl-1,3-thiazol-2-yl)morpholine

  • MDL number MFCD27981000
  • Cas Number : 1540791-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-89099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89099
Availability details for
Quantity
  • Availability
  • Add to Cart