ENA423210801 Enamine

1-amino-2-ethenylcyclopropane-1-carbonitrile

  • MDL number MFCD21262561
  • Cas Number : 1483826-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93188
Availability details for
Quantity
  • Availability
  • Add to Cart