ENA423210818 Enamine

3-(pyrimidin-2-yl)propanoic acid

  • MDL number MFCD00223696
  • Cas Number : 439108-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-180
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97262
Availability details for
Quantity
  • Availability
  • Add to Cart