ENA423210819 Enamine

2-ethyl-2-methylbut-3-enoic acid

  • MDL number MFCD19229154
  • Cas Number : 133900-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97330

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97330
Availability details for
Quantity
  • Availability
  • Add to Cart