ENA423210823 Enamine

6-bromo-4-methylpyridin-3-ol

  • MDL number MFCD18255424
  • Cas Number : 1256824-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98304
Availability details for
Quantity
  • Availability
  • Add to Cart