ENA424044845 Enamine

1-(3,5-dimethylbenzoyl)piperazine hydrochloride

  • MDL number MFCD28012173
  • Cas Number : 1803604-33-2
  • Molecular Weight : 254.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170353
Availability details for
Quantity
  • Availability
  • Add to Cart