ENA424044851 Enamine

2,3-dihydro-1H-1-benzazepine hydrochloride

  • MDL number MFCD28012183
  • Cas Number : 1803601-46-8
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-170713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170713
Availability details for
Quantity
  • Availability
  • Add to Cart