ENA424044890 Enamine

methyl 2-methylpyrrolidine-3-carboxylate hydrochloride

  • MDL number MFCD28012242
  • Cas Number : 1803567-37-4
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-methylpyrrolidine-3-carboxylate hydrochloride
Inchi Code 1S/C7H13NO2.ClH/c1-5-6(3-4-8-5)7(9)10-2;/h5-6,8H,3-4H2,1-2H3;1H
Inchi Key LCNSRHSQQQHRIA-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-172357

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-172357
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart