ENA424044907 Enamine

tert-butyl 3-amino-4-cyano-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD28012270
  • Cas Number : 1803581-55-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173647
Availability details for
Quantity
  • Availability
  • Add to Cart