ENA424044963 Enamine

3-bromo-2,4-dimethylbenzoyl chloride

  • MDL number MFCD28012348
  • Cas Number : 1803605-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174748
Availability details for
Quantity
  • Availability
  • Add to Cart