ENA424044984 Enamine

(4-methyl-3-nitrophenyl)hydrazine hydrochloride

  • MDL number MFCD28012376
  • Cas Number : 104446-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175091
Availability details for
Quantity
  • Availability
  • Add to Cart