ENA424044995 Enamine

{4-[3-(pyridin-4-yl)-1,2,4-oxadiazol-5-yl]phenyl}methanamine hydrochloride

  • MDL number MFCD28012389
  • Cas Number : 1803587-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175271
Availability details for
Quantity
  • Availability
  • Add to Cart