ENA424045018 Enamine

4-[5-(2-methoxyphenyl)-1,2,4-oxadiazol-3-yl]piperidine hydrochloride

  • MDL number MFCD28012412
  • Cas Number : 1787896-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175718
Availability details for
Quantity
  • Availability
  • Add to Cart