ENA424045058 Enamine

2-(2-chloro-6-methylphenyl)acetonitrile

  • MDL number MFCD00019828
  • Cas Number : 76574-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170809
Availability details for
Quantity
  • Availability
  • Add to Cart