ENA424045059 Enamine

(2-chloro-6-methylphenyl)methanol

  • MDL number MFCD06203023
  • Cas Number : 77206-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170810
Availability details for
Quantity
  • Availability
  • Add to Cart