ENA424045126 Enamine

1-(cyclopropylmethyl)-1H-pyrazol-4-ol

  • MDL number MFCD28012229
  • Cas Number : 1597876-36-2
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-172105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172105
Availability details for
Quantity
  • Availability
  • Add to Cart