ENA424045189 Enamine

5-methyl-6-(trifluoromethyl)pyridin-3-amine

  • MDL number MFCD11848227
  • Cas Number : 941606-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-173080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173080
Availability details for
Quantity
  • Availability
  • Add to Cart