ENA424045218 Enamine

2-[(4-methylcyclohexyl)amino]ethan-1-ol

  • MDL number MFCD11152723
  • Cas Number : 108666-82-6
  • Molecular Weight : 157.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173446
Availability details for
Quantity
  • Availability
  • Add to Cart