ENA424045222 Enamine

(3-bromo-4-iodophenyl)methanol

  • MDL number MFCD09836677
  • Cas Number : 249647-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173475
Availability details for
Quantity
  • Availability
  • Add to Cart