ENA424045305 Enamine

1-ethyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD14620003
  • Cas Number : 89852-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173865
Availability details for
Quantity
  • Availability
  • Add to Cart