ENA424045327 Enamine

1-ethoxy-3-methylbutan-2-amine

  • MDL number MFCD06247362
  • Cas Number : 883538-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-173965
Availability details for
Quantity
  • Availability
  • Add to Cart