ENA424045389 Enamine

4-chloro-6-(dimethylamino)-2-methylpyrimidine-5-carbaldehyde

  • MDL number MFCD23976600
  • Cas Number : 1508628-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174210
Availability details for
Quantity
  • Availability
  • Add to Cart