ENA424045391 Enamine

4-benzylpiperidin-2-one

  • MDL number MFCD01631136
  • Cas Number : 179683-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174212
Availability details for
Quantity
  • Availability
  • Add to Cart