ENA424045423 Enamine

2-amino-6-chloropyridin-3-ol

  • MDL number MFCD13189893
  • Cas Number : 159309-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-174371
Availability details for
Quantity
  • Availability
  • Add to Cart